missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoethanolamine solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
51.20 € - 107.00 €
Chemical Identifiers
CAS | 141-43-5 |
---|---|
Molecular Formula | C2H7NO |
Molecular Weight (g/mol) | 61.08 |
MDL Number | MFCD00008183 |
InChI Key | HZAXFHJVJLSVMW-UHFFFAOYSA-N |
PubChem CID | 700 |
ChEBI | CHEBI:16000 |
IUPAC Name | 2-aminoethanol |
SMILES | NCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12912155
|
Fisher Chemical
A 028 100 |
100 mL | Bottle |
51.20 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12922155
|
Fisher Chemical
A 028 250 |
250 mL | Bottle |
73.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12932155
|
Fisher Chemical
A 028 500 |
500 mL | Bottle |
107.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
141-43-5 | |
61.08 | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
CHEBI:16000 | |
NCCO |
C2H7NO | |
MFCD00008183 | |
700 | |
2-aminoethanol |
Specifications
1g/L | |
Single-component Standard Solution | |
MFCD00008183 | |
NCCO | |
61.08 | |
CHEBI:16000 | |
Liquid |
141-43-5 | |
C2H7NO | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
2-aminoethanol | |
700 | |
H2O | |
Monoethanolamine |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)