missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdic Acid, Honeywell Fluka™
ACS Reagent,≥85.0% MoO3 basis
84.67 € - 330.76 €
Chemical Identifiers
CAS | 7782-91-4 |
---|---|
Molecular Formula | H2MoO4 |
Molecular Weight (g/mol) | 161.96 |
MDL Number | MFCD00036287 |
InChI Key | VLAPMBHFAWRUQP-UHFFFAOYSA-L |
Synonym | molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 |
PubChem CID | 82208 |
ChEBI | CHEBI:25371 |
IUPAC Name | dihydroxy(dioxo)molybdenum |
SMILES | O[Mo](O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15672300
|
Honeywell-Fluka
232084-100G |
100 g |
84.67 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15682300
|
Honeywell-Fluka
232084-500G |
500 g |
330.76 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7782-91-4 | |
161.96 | |
VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
82208 | |
dihydroxy(dioxo)molybdenum |
H2MoO4 | |
MFCD00036287 | |
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 | |
CHEBI:25371 | |
O[Mo](O)(=O)=O |
Specifications
7782-91-4 | |
MFCD00036287 | |
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 | |
O[Mo](O)(=O)=O | |
161.96 | |
CHEBI:25371 | |
ACS Reagent | |
Molybdic Acid |
H2MoO4 | |
NONH for all modes of transport | |
VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
dihydroxy(dioxo)molybdenum | |
82208 | |
≥85% (MoO3 basis) | |
3.1g/mL at 25°C; Vapor Density: 6.2 (vs air) |
Safety and Handling
P261-P305 + P351 + P338
H319-H335-H373
missing translation for 'einecsNumber' : 231-970-5