missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, 99.5%, Thermo Scientific Chemicals

Product Code. 11469083
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in the oil industry. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in the oil industry. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.

Solubility
Slightly soluble in water. Soluble in concentrated mineral acids and alkali hydroxide solutions.

Notes
It does not react with acids other than hydrofluoric and concentrated sulfuric acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Boiling Point 1155°C (sublimation)
Melting Point 795°C
Quantity 1000 g
UN Number UN3288
Merck Index 14,6239
Solubility Information Slightly soluble in water. Soluble in concentrated mineral acids and alkali hydroxide solutions.
Formula Weight 143.94
Percent Purity 99.5%
Odor Odorless
Density 4.692
Chemical Name or Material Molybdenum (VI) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'