Learn More
Molybdenum(V) isopropoxide, 99.6% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals
173.00 € - 491.00 €
Chemical Identifiers
CAS | 209733-38-0 |
---|---|
Molecular Formula | C15H35MoO5 |
Molecular Weight (g/mol) | 391.39 |
MDL Number | MFCD00210636 |
InChI Key | HLFWWCCBFJUYJL-UHFFFAOYSA-N |
Synonym | molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate |
PubChem CID | 18475294 |
IUPAC Name | molybdenum;propan-2-olate |
SMILES | CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363528
|
Thermo Scientific Alfa Aesar
039159.AC |
25 mL |
173.00 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11373528
|
Thermo Scientific Alfa Aesar
039159.AE |
100 mL |
491.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Molybdenum(V) isopropoxide is used to prepare novel MoVSbOxtype catalysts for selective isobutane oxidation. It is used to prepare molybdenum Oxide Nanofibers by electrospinning.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
209733-38-0 | |
391.39 | |
HLFWWCCBFJUYJL-UHFFFAOYSA-N | |
18475294 | |
CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C |
C15H35MoO5 | |
MFCD00210636 | |
molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate | |
molybdenum;propan-2-olate |
Specifications
209733-38-0 | |
12°C (54°F) | |
C15H35MoO5 | |
MFCD00210636 | |
Moisture sensitive | |
Miscible with alcohols,ketones and esters. | |
CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C | |
391.39 | |
391.04 | |
5% w/v in isopropanol | |
(metals basis) |
Yellow to Brown | |
Alcohol-like | |
Mo[OCH(CH3)2]5 | |
UN1219 | |
molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate | |
HLFWWCCBFJUYJL-UHFFFAOYSA-N | |
molybdenum;propan-2-olate | |
18475294 | |
99.6% | |
Liquid | |
Molybdenum(V) isopropoxide |
Safety and Handling
GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H225-H319-H335-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only