missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum rod, 6.4mm (0.25in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11366727
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Approx. 15.2cm/50g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Color Gray
Appearance Dark gray rod
Assay Percent Range 99.95% (Metals basis)
Quantity 250 g
Odor Odorless
Form Rod
Chemical Name or Material Molybdenum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'