Learn More
Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads, Thermo Scientific Chemicals
46.43 € - 122.61 €
Chemical Identifiers
CAS | 69912-79-4 |
---|---|
Molecular Formula | AlNaO6Si2 |
Molecular Weight (g/mol) | 202.14 |
MDL Number | MFCD00147627 |
InChI Key | URGAHOPLAPQHLN-UHFFFAOYSA-N |
PubChem CID | 19758701 |
IUPAC Name | aluminum;sodium;dioxido(oxo)silane |
SMILES | [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327575
|
Thermo Scientific Alfa Aesar
L05722.30 |
250 g |
46.43 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337575
|
Thermo Scientific Alfa Aesar
L05722.0B |
1000 g |
122.61 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Molecular sieves, 13X, 3-5mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69912-79-4 | |
202.14 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane |
AlNaO6Si2 | |
MFCD00147627 | |
19758701 | |
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O |
Specifications
Hygroscopic | |
AlNaO6Si2 | |
Insoluble in water. | |
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O | |
202.14 | |
Molecular sieves, 5A, 3 to 5mm (0.12 to 0.20 in) beads |
69912-79-4 | |
MFCD00147627 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane | |
19758701 |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only