missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 4A, Powder, Activated, −325 mesh, Honeywell Fluka™

Product Code. 15646710
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Powder, activated, −325 mesh particle size

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Color Tan
Density 2g/cm3
Packaging Poly bottle
Quantity 25 g
Physical Form Powder, activated
Melting Point 400°C
Linear Formula Na12[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
Chemical Name or Material Molecular sieves, 4A
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'