missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 4A, Powder, Activated, −325 mesh, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Color | Tan |
Density | 2 g/cm3 |
Packaging | Poly bottle |
Quantity | 1 kg |
Physical Form | Powder, activated |
Melting Point | 400°C |
Linear Formula | Na12[(AlO2)12(SiO2)12] · xH2O |
UN Number | NONH for all modes of transport |
IUPAC Name | (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid |
Chemical Name or Material | Molecular sieves, 4A |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'