missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 4 Å, 8 to 12mesh, Honeywell Fluka™
Beads, 8-12 mesh
50.28 € - 748.20 €
Chemical Identifiers
CAS | 70955-01-0 |
---|---|
Molecular Formula | C20H25FN2O8 |
Molecular Weight (g/mol) | 440.42 |
MDL Number | MFCD00131613 |
InChI Key | FJUZEZRZKVMAMD-UHFFFAOYNA-N |
PubChem CID | 73906282 |
IUPAC Name | (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid |
SMILES | COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15641850
|
Honeywell-Fluka
208604-250G |
250 g | N/A | ||||||
15631850
|
Honeywell-Fluka
208604-1KG |
1 kg |
215.76 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15651850
|
Honeywell-Fluka
208604-5KG |
5 kg |
748.20 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
70955-01-0 | |
440.42 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid |
C20H25FN2O8 | |
MFCD00131613 | |
73906282 | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Specifications
8 to 12mesh | |
C20H25FN2O8 | |
MFCD00131613 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid | |
73906282 |
70955-01-0 | |
Na12[(AlO2)12(SiO2)12] · xH2O | |
NONH for all modes of transport | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC | |
440.42 | |
Molecular Sieves, 4 Å |
Safety and Handling
P261-P305 + P351 + P338
H315-H319-H335