missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular sieves 3A, powder <50 micron, Thermo Scientific Chemicals
69.75 € - 646.00 €
Chemical Identifiers
CAS | 308080-99-1 |
---|---|
Molecular Formula | AlNaO6Si2 |
Molecular Weight (g/mol) | 202.135 |
MDL Number | MFCD00131613 |
InChI Key | URGAHOPLAPQHLN-UHFFFAOYSA-N |
PubChem CID | 19758701 |
IUPAC Name | aluminum;sodium;dioxido(oxo)silane |
SMILES | [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10072690
|
Thermo Scientific Acros
214795000 |
500 g | Glass bottle |
69.75 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10249163
|
Thermo Scientific Acros
214790025 |
2.5 kg | Plastic Drum |
256.28 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15119243
|
Thermo Scientific Acros
214790100 |
10 kg | Plastic drum | N/A | N/A | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agentChemical Identifiers
308080-99-1 | |
202.135 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane |
AlNaO6Si2 | |
MFCD00131613 | |
19758701 | |
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
Specifications
Undesignated | |
500 Micron Max. | |
> 22% (50%rH, 20°C, 24h) | |
Powder or Crystals | |
MFCD00131613 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane | |
19758701 |
11.4 (10% max. aq. suspension) | |
< 1.5% (550°C, 2 hrs.) | |
308080-99-1 | |
AlNaO6Si2 | |
Solubility in water: insoluble. | |
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] | |
202.135 | |
Molecular sieves 3A |
Safety and Handling
HYGROSCOPIC
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Molecular sieves 3A, powder <50 micron, Thermo Scientific Chemicals