missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves 3A, Pellets, Dia.: 3.2mm, Honeywell Fluka™

Product Code. 15623960
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with inliner
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pellets, 3.2 mm

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

Color Beige or Tan
Density 2 g/cm3
Packaging Drum with inliner
Particle Size Dia.: 3.2 mm
Quantity 5 kg
Physical Form Pellets
Melting Point >400°C
Linear Formula KnNa12-n[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
IUPAC Name aluminum;sodium;dioxido(oxo)silane
Chemical Name or Material Molecular sieves, UOP Type 3Å
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'