Learn More
Invitrogen™ Low Density Lipoprotein from Human Plasma, BODIPY™ FL complex (BODIPY™ FL LDL)
Nonacetylated conjugate
Brand: Invitrogen™ L3483
521.35 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Human low-density lipoprotein (LDL) is large protein complex (approx. 500,000 Da) that binds to a specific receptor on the surface of vertebrate cells and delivers cholesterol via receptor-mediated endocytosis. Experiments to study LDL are typically performed by adding fluorescently labeled LDL to cultured cells and analyzing them by microscopy or flow cytometry. Alternatively the fluorescently labeled LDL can be injected into test animals, and the distribution of label can be analyzed after specified time period.
- Label (Ex/Em): BODIPY FL (515/520)
- Acetylated: No
- Amount: 200μL (1.0mg/mL)
- Prepared from fresh human plasma approximately every two months
- Key Applications: Counting cell-surface LDL receptors, and analyzing their motion and clustering following internalization; Quantitating LDL receptor activity in fibroblasts (replacing the radiolabeled LDL assay); Investigating LDL expression and identifying LDL receptor deficiencies in various cell lines
Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles
Order Info
Shipping Condition: Wet Ice
Specifications
1 mg⁄ml | |
Fluorescence | |
BODIPY Dyes | |
BODIPY™ | |
Wet Ice |
Store in refrigerator (2–8°C) and protect from light. | |
Supplement | |
Liquid | |
200 μL |
For Research Use Only. Not for use in diagnostic procedures.