Learn More
Invitrogen™ Low Density Lipoprotein from Human Plasma, Acetylated, DiI complex (DiI AcLDL)
Acetylated (Ac) apoprotein used to study cell types that express receptors specific for this acetylated version, i.e., endothelial and microglial cells
Brand: Invitrogen™ L3484
653.60 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Human low-density lipoprotein (LDL) is large protein complex (approx. 500,000 Da) that binds to a specific receptor on surface of vertebrate cells and delivers cholesterol via receptor-mediated endocytosis. Experiments to study LDL are typically performed by adding fluorescently labeled LDL to cultured cells and analyzing them by microscopy or flow cytometry. Alternatively the fluorescently labeled LDL can be injected into test animals, and the distribution of the label can be analyzed after the specified time period.
- Acetylated: Yes
- Amount: 200μL (1.0mg/mL)
- LDL and AcLDL products from fresh human plasma approximately every two months and are available on continuous basis
- LDL containing an unmodified apoprotein is used to study normal cholesterol delivery and internalization: If lysine residues of LDL apoprotein have been acetylated, the LDL complex no longer binds to the LDL receptor, but rather is taken up by endothelial and microglial cells that possess ;scavenger receptors specific for that modified form
- Some of many applications for labeled LDL complexes include: Counting cell-surface LDL receptors, and analyzing their motion and clustering following internalization, Quantitating LDL receptor activity in fibroblasts (replacing the radiolabeled LDL assay), Investigating LDL expression and identifying LDL receptor deficiencies in various cell lines
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Wet Ice
Specifications
1 mg⁄ml | |
Fluorescence | |
Other Label(s) or Dye(s) | |
6 to 8 | |
Wet Ice |
Store in refrigerator (2–8°C) and protect from light. | |
Supplement | |
Liquid | |
200 μL |
For Research Use Only. Not for use in diagnostic procedures.