missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Live Cell Imaging Solution
GREENER_CHOICE

Physiological medium developed for live cell imaging applications that has broad utility in cell-based research, including dye loading and wash steps

Brand:  Invitrogen™ A14291DJ

Product Code. 12363603

This item is not returnable. View return policy

Description

Description

Live cell imaging can be challenging. Not only must staining be optimized for particular assay readout, spectral compatibility, and signal-to-noise level, but live cell imaging of dynamic processes and cell physiology requires live observation over time. The choice of medium is particularly important for time-lapse imaging and experiments where cells are exposed to ambient conditions for longer periods. For reliable results, it is essential that cells be healthy and maintained as closely as possible to physiological temperature, pH, oxygen tension, and other conditions. If cells appear unhealthy they should not be used for imaging.

  • Optically clear solution buffered with HEPES at pH 7.4 that keeps cells healthy for up to 4 hours at ambient atmosphere and temperature
  • Suitable solution for live cell fluorescence imaging applications
  • Extended buffering capacity compared to media for improved cell viability
  • Developed specifically for live cell imaging applications: provides image clarity, signal-to-noise ratio, and cell viability that are significantly enhanced compared to standard media

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Sample Preparation

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Live Cell Imaging Solution
Physiological Imaging Buffer
Sustainable packaging
Room Temperature
Molecular Probes
Store at ≤25 °C
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Live Cell Imaging Solution > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.