Learn More
Invitrogen™ Staphylococcus aureus (Wood strain without protein A) BioParticles™, unlabeled
Unlabeled, heat- or chemically killed bacteria and yeast in a variety of sizes, shapes, and natural antigenicities
Brand: Invitrogen™ S2859
127.33 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
- Useful in phagocytosis assays, both as controls and for custom Labeling with reactive dyes
- Label (Ex/Em): Unlabeled
- Particle: S. aureus (Wood strain, without protein A)
- Provided as lyophilized powders
- Approximately 3 x 108 S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid
- Conjugates can be reconstituted in the buffer of choice for use in phagocytosis assays
Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Flow Cytometry, Flow Cytometry Endocytosis, Exocytosis and Phagocytosis Assays, Flow Cytometry of Cellular Processes
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C). | |
Fluorescence Microscope | |
Unconjugated | |
BioParticles™ | |
Room Temperature |
Fluorescence | |
Bioparticle Unlabeled | |
Lyophilized Powder | |
100 mg | |
S. aureus |
For Research Use Only. Not for use in diagnostic procedures.