Learn More
Invitrogen™ Fura Red™, AM, cell permeant
Visible light-excitable fura-2 analog that offers particular possibilities for ratiometric measurement of Ca2+ in single cells
750.00 € - 798.00 €
Specifications
Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
---|---|
Detection Method | Fluorescence |
For Use With (Application) | Cell Viability and Proliferation |
For Use With (Equipment) | Fluorescence Microscope, Microphotometer, Flow Cytometer |
Dye Type | Fluorescent Dye-Based |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11598956
|
Invitrogen™
F3020 |
500 μg |
750.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11508966
|
Invitrogen™
F3021 |
10 x 50μg |
798.00 €
500µg |
In Stock
Log in to see stock. |
|||||
Description
- Measure by microphotometry, imaging or flow cytometry when used with single excitation, green-fluorescent calcium indicator
- Acetoxymethyl (AM) ester form
- Useful for noninvasive intracellular loading
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Cell-Based Second Messenger Assays, Drug Discovery and Development, G-Protein Coupled Receptor Biology, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Cell Viability and Proliferation | |
Fluorescent Dye-Based | |
Room Temperature |
Fluorescence | |
Fluorescence Microscope, Microphotometer, Flow Cytometer | |
Fura Red™ |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.