Learn More
Thermo Scientific™ Molecular BioProducts™ PCR Tubes and Plates
Molded from 100% virgin polypropylene
Specifications
Material Polypropylene
View More Specs
Description
Description
Molded from 100% virgin polypropylene
Tubes feature thin walls for optimal heat transfer and caps that seal securely to minimize evaporation. Plates feature elevated well rims to ensure uniform sealing.
  • PurePak™ packaging reduces contamination from reaching into a bulk bag of tubes by dividing tubes into 10 separate PurePaks
  • Certified RNase-, DNase-, DNA, and Pyrogen-free
  • Clear, nonsterile
Specifications
Specifications
Polypropylene
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Molecular BioProducts™ PCR Tubes and Plates