Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
Brand:  Epredia™ 88027
 View more versions of this product
Product Code. 12593647
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green
Specifications
Specifications
Periodic Acid Schiff
125mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : periodic acid (H5IO6)