missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit

Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)

Brand:  Epredia™ 88036


 View more versions of this product

Product Code. 12563657

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green
Specifications

Specifications

Silver Nitrate Solution (5%)
30mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Carcinogenicity Category 1B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P363-Wash contaminated clothing before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : silver nitrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers