Learn More
Hamilton™ Thermo Finnagan™ Cemented Needle Syringes
Enables the automatic introduction of samples into the injector of the gas chromatograph.
Brand:  Hamilton™ 204000
Product Code. 11547614
  • 130.00 € / Each
Explore more special offers
Description
Description
  • Improves reproducibility and speed
  • Point style specifically designed to withstand repeated penetration through the GC septa
  • For use with Thermo Finnigan™ AI/AS 3000 TriPlus™ autosamplers