Learn More
Miele Professional™ Mobile unit
Mobile unit E 940 for the optimum loading of narrow necked glassware on 2 levels
Brand:  Miele Professional™ 4607630
Product Code. 15450555
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • The mobile unit can also be used without the upper level
  • Magnetic strip for automatic mobile unit recognition
  • Lower level equipped with a total of 115 nozzles with clips
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel, Plastic
Stainless Steel, Black
Washer-disinfectors, laboratory
565 mm
Mobile Unit for Mesh Trays
115 Narrow-necked glassware
790 mm
640 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Miele Professional™ Mobile unit > Dimensions (D x W X H): 790 x 640 x 565mm