missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Mobile pedestal, oak two drawer, 600mm x 410mm x 600mm
Brand: Manutan 112D014
This item is not returnable.
View return policy
Description
Scratch-resistant melamine faced chipboard. Accepts A4 or foolscap, foolscap files are stored lengthways and A4 files are stored width ways. Easy self-assembly.- Blonde oak finish
- Two drawer
- Dimensions (H x W x D) : 600mm x 410mm x 600mm