Learn More
MNS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12693-1-AP-150UL
 View more versions of this product
Product Code. 16800554
  • 410.00 € / 150µL
Explore more special offers
Description
Description
MNS1 may play a role in the control of meiotic division and germ cell differentiation through regulation of pairing and recombination during meiosis.
Specifications
Specifications
MNS1
Polyclonal
Unconjugated
MNS1
meiosis nuclear structural 1, MNS1
Rabbit
Antigen Affinity Chromatography
RUO
17427, 363093, 55329
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q61884, Q6AXQ8, Q8NEH6
Mns1
MNS1 Fusion Protein Ag3423
150 μL
Primary
Human, Mouse, Rat