missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ MicroSeal Supersorb™ Flat Mop Wipers

MicroSeal SuperSorb™ is a sealed edge cleanroom laundered 100% polyester knit wiper.

Brand:  Berkshire™ MSSS.0824.12

 View more versions of this product

Product Code. 17812135

  • 1131.71 € / Pack of 300

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Critically low particles, fibers, ions and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, Acetone and other solvents
  • Highest sorbency level available in a polyester knit reduces wiper usage
  • Pinsonic design adds surface texture that aids in stubborn residue removal
  • Low cost in use compared to other two-ply polyester knits
Specifications

Specifications

8 x 24 in. (20 x 61 cm)
White
Non-sterile
Autoclavable
24 in.
For ISO Class 4 (Class 10) cleanrooms application
Smooth Polyester
Flat Head Mop Cover
8 in.
20 cm
61 cm
Single-use
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers