Learn More
OHAUS™ Microplate Holder
Designed to hold standard microplate
Brand:  OHAUS™ 30400213
 View more versions of this product
Product Code. 15752629
  • 71.75 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Ohaus Microplate Vortex Mixer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Two standard microplates
Plasitc
Re-Suspending Liquids, Mixing Reagents, Buffers and Emulsions, ELISA Assays, Drug Extractions, Mixing Samples, PCR, Digestions, Cell Disruptions, Genotyping
1 Ea.
Double Microplate Holder
Double Microplate Holder
Microplate Vortex Mixer/Heavy Duty Vortex Mixer