Learn More
Ohaus™ Microplate Clamp
OHAUS™ Microplate Clamp designed to securely hold a microplate in place.
Brand: Ohaus™ 30400104
This item is not returnable.
View return policy
Description
- OHAUS offers a range of shaker accessories to meet specific sample requirements
- A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
- Platforms can increase the amount of usable space for samples
- In some cases can be stacked to increase throughput and double sample capacity
- Tube racks and dimpled mats securely hold tubes in place
Specifications
Clamp | |
Stainless Steel | |
Microplates | |
Clamp Microplate Stainless Steel |