Learn More
Micronova™ T-Mop™ PolyTex Cover
Low profile mop with a wide surface serves as a perfect device for applying disinfectants in a sterile environment
Brand: Micronova™ T712-18
This item is not returnable.
View return policy
Description
- Constructed textured polyester over foam slipcovers
- 18 in. (45.72cm) in size
- Light weight, economical enough for single use and autoclavable
- With a 180° swivel, the mop is ideal for cleaning under equipment or for applying disinfectants or detergents to large surface areas
- Smaller adapter heads can be used for cleaning surfaces and awkward angles on shelving and in narrow chambers
Specifications
Mop Cover | |
Reusable | |
Polytex |
Autoclavable | |
Ideal for cleaning under equipment or for applying disinfectants or detergents to large surface areas |