missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ Microlink Cap
Shop All Wheaton Science Products Products
Click to view available options
Closure Size:
20-400 mm
33-400 mm
Specifications
Specifications
Color | White |
For Use With (Application) | Prolonged Period of Time Over Wide Temperature Range |
Autoclavable | Autoclavable |
Product Type | Microlink Cap |
Closure Size | 33-400 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction