Learn More
Sigma Laborzentrifugen™ Microhematocrit Rotor Reader
For determining the ratio of the volume of packed red blood cells to the volume of whole blood by centrifuging.
Specifications
Microhematocrit Rotor Reader |