Learn More
Ansell™ Microgard™ 2300 Plus Coveralls with Hood
Product Code. 15526614
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Provides an excellent barrier to many harmful chemicals while being lightweight and yet relatively strong and durable. Ansell Microgard 2300 Plus is designed for workers involved in environmental clean-up, general industrial and chemical handling applications.
  • Category III 3-piece hood
  • Elasticated hood, finger loops, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
  • Polycoated bi-component barrier
  • Highly visible bright yellow color improves worker safety
  • Lightweight yet relatively strong and durable
  • Anti-static according to EN 1149-5
  • Includes a respirator fit hood and a zip flap with self-adhesive tape closure
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size 2X-Large
Material Polyethylene-coated Non-woven Bicomponent Fabric
Color Yellow
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Zipper with Storm Flap
Seam Type Stitched, Taped
Coating Polyethylene
Certifications/Compliance Type 3-B, Type 4-B, Type 5-B, EN 1073-2, EN 14126, EN 1149-5
Description 2-way front zip with resealable storm flap
No. of Pockets None
Style Includes Hood, finger loops