Learn More
Ansell Microgard™ 1500 Plus White Coveralls with Hood
Product Code. 15536494
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Provides a good barrier against particulates such as asbestos, brick dust and cement dust. Ansell Microgard™ 1500 Plus White Coveralls with Hood filter 99.9% of particles >3μ.
  • Coverall with 3-piece hood, stitched seams, elasticated hood, waist, wrists and ankles
  • 2-way front zipper with re-sealable stormflap
  • Anti-static treated SMS fabric
  • Breathable to help reduce the risk of heat stress
  • Silicone-free which is critical in spray painting applications
  • Optimized body fit for wearer comfort and safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Anti-static SMS
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Certifications/Compliance (Europe) EN ISO 13982-1, EN13034, EN1073-2, EN1149-5
Model WH15-S-00-111
No. of Pockets None
Style Includes Hood