Learn More
Ansell™ Microchem™ 4000 Coveralls with Hood and Integrated Socks
Product Code. 15526454
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Designed to provide an exceptional barrier against organic and inorganic chemicals and biological agents. A unique multi-layer barrier fabric renowned for its lightweight, yet robust textile feel and exceptional barrier to organic and inorganic chemicals.
  • Coverall with elasticated 2-piece hood with integrated socks and boot overflap
  • Front entry double-zip system.
  • Elasticated double cuffs (knitted inner cuff), waist and ankles.
  • Permeation tested against over 180 chemicals, including chemical warfare agents
  • Textile-like inner improves wearer acceptance
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size 2X-Large
Material Multi-layer Barrier Laminate Fabric
Color Green
Waist Style Elastic
Ankle Style Elastic, Attached Socks, Boot Overflaps
Cuff Style Double, Elastic
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper Front Entry
Seam Type Taped, Welded
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5
Description Hood, integrated socks with boot overflap
No. of Pockets None
Style Includes Hood, integrated socks with boot overflap