missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Microcentrifuge Tubes with Screw Cap

The cover thread (PP) and reaction tube (PP) are perfectly matched, and in combination with the silicone seal offer reliable protection against liquid leakage or penetration

Brand:  BRAND™ 780759

266.44 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.


 View more versions of this product

Product Code. 10403675

  • 326.00 € / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sterile reaction tubes are delivered with attached  screw-on covers and are non-mutagenic and non-cytotoxic
  • With matte label field
  • Application: -196 °C to 121 °C
  • sterility/SAL (Sterility Assurance Level) 10-6
  • Free from DNA, DNase, RNase, endotoxins
  • Colored covers are available for efficient sample management
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    13 mm
    20,000 G
    Polypropylene
    Without base
    Screw Cap
    Up to 1.2 mL
    2 mL
    44 mm
    Yes
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers