missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGEA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14711-1-AP-150UL
This item is not returnable.
View return policy
Description
Two bands at 130 kDa and 75 kDa could be detected using antibody 66033-1-Ig.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predomitly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.Specifications
MGEA5 | |
Polyclonal | |
Unconjugated | |
MGEA5 | |
Beta N acetylhexosaminidase, Bifunctional protein NCOAT, HAT, HEXC, Hexosaminidase C, KIAA0679, MEA5, Meningioma expressed antigen 5, MGEA5, N acetyl beta glucosaminidase, NCOAT, O GlcNAcase, OGA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10724, 154968, 76055 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60502, Q8VIJ5, Q9EQQ9 | |
MGEA5 | |
MGEA5 Fusion Protein Ag6405 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction