missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFSD7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21037-1-AP-20UL
This item is not returnable.
View return policy
Specifications
MFSD7 | |
Polyclonal | |
Unconjugated | |
MFSD7 | |
LP2561, MFSD7, MYL5, UNQ385/PRO717 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
243197, 305625, 84179 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UXD7, Q8CE47 | |
MFSD7 | |
MFSD7 Fusion Protein Ag15361 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction