Learn More
MFSD7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21037-1-AP-150UL
 View more versions of this product
Product Code. 16800635
  • 410.00 € / 150µL
Explore more special offers
Specifications
MFSD7
Polyclonal
Unconjugated
MFSD7
LP2561, MFSD7, MYL5, UNQ385/PRO717
Rabbit
Antigen Affinity Chromatography
RUO
243197, 305625, 84179
-20°C
Liquid
Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6UXD7, Q8CE47
MFSD7
MFSD7 Fusion Protein Ag15361
150 μL
Primary
Human, Mouse, Rat