missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFSD10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11518-1-AP-20UL
This item is not returnable.
View return policy
Description
TETRAN′s overexpression in cultured human cells caused resistance to some NSAIDs (Non-steroidal anti-inflammatory drugs), suggesting that TETRAN is an efflux pump for some NSAIDs.Specifications
MFSD10 | |
Polyclonal | |
Unconjugated | |
MFSD10 | |
MFSD10, TETRAN, TETTRAN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10227, 305449, 68294 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14728, Q9D2V8 | |
Mfsd10 | |
MFSD10 Fusion Protein Ag2080 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction