Learn More
MFN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12186-1-AP-20UL
 View more versions of this product
Product Code. 16879743
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This gene encodes a mitochondrial membrane protein that participates in mitochondrial fusion and contributes to the maintece and operation of the mitochondrial network. This protein is involved in the regulation of vascular smooth muscle cell proliferation, and it may play a role in the pathophysiology of obesity. Mutations in this gene cause Charcot-Marie-Tooth disease type 2A2, and hereditary motor and sensory neuropathy VI, which are both disorders of the peripheral nervous system. Defects in this gene have also been associated with early-onset stroke. Two transcript variants encoding the same protein have been identified.
Specifications
Specifications
MFN2
Polyclonal
Unconjugated
MFN2
CMT2A, CMT2A2, CPRP1, HSG, KIAA0214, MARF, MFN2, Mitofusin 2, Transmembrane GTPase MFN2
Rabbit
Antigen Affinity Chromatography
RUO
170731, 64476, 9927
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95140, Q80U63, Q8R500
Mfn2
MFN2 Fusion Protein Ag2845
20 μL
Primary
Rat, Human, Mouse