missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Lab Animal Feed (LAR) Cart

Keep feeding efforts to a minimum. Metro Lab Animal Feed Cart is a two-shelf unit on casters making feeding laboratory animals more convenient for staff.

Brand:  Metro™ LAR2

 View more versions of this product

Product Code. 15966971

  • 1190.00 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Super Erecta™ shelf construction provides easy assembly and absolute rigidity
  • Relocation of shelves is quick and simple
  • Stainless steel finish provides superior corrosion resistance
  • Open shelf design minimizes dust accumulation while maximizing air circulation and visibility
  • Compact footprint for easy maneuvering in tight lab spaces

Specifications

Specifications

Heavy-duty Lab Animal Feed Cart
Designed to store and transport animal feed
99 cm
36 in.
600 lb.
Stainless Steel
24 in.
36 x 24 x 39 in. (91 x 61 x 99 cm)
39 in.
Two totes and one shelf
91 cm
273 kg
1
61 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.