Learn More
Emma Safety Footwear Metric XD Safety Shoes
Unique microfiber upper is fully washable. Features SmartecAir™ for dry, fresh feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors often seen in high-tech and cleanroom environments.
Brand:  Emma Safety Footwear MM50756044
 View more versions of this product
Product Code. 15908631
  • 110.24 € / 1 pair
Estimated Shipment: 17-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
Metric XD
Microfiber
44