missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylurea, 97%, Thermo Scientific Chemicals
22.71 € - 74.47 €
Chemical Identifiers
CAS | 598-50-5 |
---|---|
Molecular Formula | C2H6N2O |
Molecular Weight (g/mol) | 74.08 |
MDL Number | MFCD00007950 |
InChI Key | XGEGHDBEHXKFPX-UHFFFAOYSA-N |
Synonym | 1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea |
PubChem CID | 11719 |
ChEBI | CHEBI:44383 |
IUPAC Name | methylurea |
SMILES | CNC(=O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10001360
|
Thermo Scientific Acros
127991000 |
100 g | Plastic bottle |
22.71 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10021360
|
Thermo Scientific Acros
127995000 |
500 g | Plastic bottle |
74.47 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
598-50-5 | |
74.08 | |
XGEGHDBEHXKFPX-UHFFFAOYSA-N | |
11719 | |
methylurea |
C2H6N2O | |
MFCD00007950 | |
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea | |
CHEBI:44383 | |
CNC(=O)N |
Specifications
598-50-5 | |
100.0 | |
White | |
96% min. (on dry substance) (HPLC) | |
CH3NHCONH2 | |
04, 64 | |
Solubility in water: 1000g/L (20°C). Other solubilities: freely soluble in ethanol | |
CNC(=O)N | |
74.08 | |
CHEBI:44383 | |
97% | |
Methylurea, 97% |
96.0 | |
98.0°C to 102.0°C | |
Authentic | |
C2H6N2O | |
MFCD00007950 | |
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea | |
XGEGHDBEHXKFPX-UHFFFAOYSA-N | |
methylurea | |
11719 | |
74.08 | |
Crystalline Solid |
Safety and Handling
missing translation for 'einecsNumber' : 209-935-0
missing translation for 'rtecsNumber' : YT7175000
missing translation for 'tsca' : TSCA
RUO – Research Use Only