Learn More
Methyltriphenylphosphonium Bromide, 98%, Thermo Scientific Chemicals
24.92 € - 761.79 €
Chemical Identifiers
CAS | 1779-49-3 |
---|---|
Molecular Formula | C19H18BrP |
Molecular Weight (g/mol) | 357.23 |
MDL Number | MFCD00011804 |
InChI Key | LSEFCHWGJNHZNT-UHFFFAOYSA-M |
Synonym | methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide |
PubChem CID | 74505 |
IUPAC Name | methyl(triphenyl)phosphanium;bromide |
SMILES | [Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10041850
|
Thermo Scientific Acros
156950100 |
10 g | Plastic bottle |
24.92 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10031850
|
Thermo Scientific Acros
156951000 |
100 g | Plastic bottle |
51.66 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10319083
|
Thermo Scientific Acros
156955000 |
500 g | Plastic bottle |
191.96 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11975801
|
Thermo Scientific Acros
156950025 |
2.5 kg | Plastic bottle |
761.79 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)Chemical Identifiers
1779-49-3 | |
357.23 | |
LSEFCHWGJNHZNT-UHFFFAOYSA-M | |
74505 | |
[Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C19H18BrP | |
MFCD00011804 | |
methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide | |
methyl(triphenyl)phosphanium;bromide |
Specifications
1779-49-3 | |
White | |
Authentic | |
C19H18BrP | |
MFCD00011804 | |
methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide | |
LSEFCHWGJNHZNT-UHFFFAOYSA-M | |
methyl(triphenyl)phosphanium;bromide | |
74505 | |
98% | |
Methyltriphenylphosphonium bromide |
230.0°C to 234.0°C | |
>240°C | |
97.5% min. (Argentometry) | |
CH3P(C6H5)3Br | |
01,697 | |
Solubility in water: 400g/L (25°C). Other solubilities: 950g/L methanol (20°C),soluble in aliphatic alcohols and chloroform,practically insoluble in arom. hydrocarbons and,acetone | |
[Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
357.23 | |
357.22 | |
Powder |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 217-218-9
- Methyltriphenylphosphonium bromide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.