Learn More
Methyltriethoxysilane, 98%, Thermo Scientific Chemicals
48.64 € - 157.79 €
Chemical Identifiers
CAS | 2031-67-6 |
---|---|
Molecular Formula | C7H18O3Si |
Molecular Weight (g/mol) | 178.30 |
MDL Number | MFCD00009066 |
InChI Key | CPUDPFPXCZDNGI-UHFFFAOYSA-N |
Synonym | methyltriethoxysilane, triethoxy methyl silane, methyl triethoxysilane, silane, triethoxymethyl, triethoxysilylmethane, union carbide a-162, ici-ep 5850, silane, methyltriethoxy, methaneorthosiliconic acid, triethyl ester, mtes |
PubChem CID | 16241 |
IUPAC Name | triethoxy(methyl)silane |
SMILES | CCO[Si](C)(OCC)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11326776
|
Thermo Scientific Alfa Aesar
L04059.AK |
250 mL |
48.64 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336776
|
Thermo Scientific Alfa Aesar
L04059.AU |
1000 mL |
157.79 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyltriethoxysilane is used in production of silicone polymers or silicone resin. It is used in non-metal surface treatment, non-aqueous polymer preparation, manufacture of semi-conductors, coatings and sealants. It finds application as cross linking agent and an adhesion promoter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyltriethoxysilane is used in production of silicone polymers or silicone resin. It is used in non-metal surface treatment, non-aqueous polymer preparation, manufacture of semi-conductors, coatings and sealants. It finds application as cross linking agent and an adhesion promoter.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
2031-67-6 | |
178.30 | |
CPUDPFPXCZDNGI-UHFFFAOYSA-N | |
16241 | |
CCO[Si](C)(OCC)OCC |
C7H18O3Si | |
MFCD00009066 | |
methyltriethoxysilane, triethoxy methyl silane, methyl triethoxysilane, silane, triethoxymethyl, triethoxysilylmethane, union carbide a-162, ici-ep 5850, silane, methyltriethoxy, methaneorthosiliconic acid, triethyl ester, mtes | |
triethoxy(methyl)silane |
Specifications
141°C to 143°C | |
2031-67-6 | |
CH3Si(OCH2CH3)3 | |
UN1993 | |
23°C (73°F) | |
Immiscible with water. | |
CCO[Si](C)(OCC)OCC | |
178.30 | |
178.3 | |
Mild | |
Moisture sensitive | |
Methyltriethoxysilane |
<-50°C | |
C7H18O3Si | |
MFCD00009066 | |
1742453 | |
methyltriethoxysilane, triethoxy methyl silane, methyl triethoxysilane, silane, triethoxymethyl, triethoxysilylmethane, union carbide a-162, ici-ep 5850, silane, methyltriethoxy, methaneorthosiliconic acid, triethyl ester, mtes | |
CPUDPFPXCZDNGI-UHFFFAOYSA-N | |
triethoxy(methyl)silane | |
16241 | |
98% | |
1.3835 | |
0.897 |
Safety and Handling
GHS H Statement
H226-H335
Flammable liquid and vapor.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 217-983-9
missing translation for 'rtecsNumber' : VV4640000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.