Learn More
Methylmagnesium bromide, 3.2M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
99.00 € - 452.00 €
Chemical Identifiers
CAS | 75-16-1, 96-47-9 |
---|---|
Molecular Formula | CH3BrMg |
Molecular Weight (g/mol) | 119.24 |
MDL Number | MFCD00000041 |
InChI Key | AVFUHBJCUUTGCD-UHFFFAOYSA-M |
Synonym | methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent |
PubChem CID | 6349 |
SMILES | C[Mg]Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10472805
|
Thermo Scientific Acros
391121000 |
100 mL | AcroSeal™ Glass bottle |
99.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11395811
|
Thermo Scientific Acros
391128000 |
800 mL | AcroSeal™ Glass Bottle |
452.00 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-16-1, 96-47-9 | |
119.24 | |
AVFUHBJCUUTGCD-UHFFFAOYSA-M | |
6349 |
CH3BrMg | |
MFCD00000041 | |
methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent | |
C[Mg]Br |
Specifications
75-16-1, 96-47-9 | |
Liquid | |
CH3MgBr | |
−11°C | |
AVFUHBJCUUTGCD-UHFFFAOYSA-M | |
bromo(methyl)magnesium | |
6349 | |
34 to 36% |
Brown | |
CH3BrMg | |
MFCD00000041 | |
methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent | |
C[Mg]Br | |
119.24 | |
119.24 | |
Methylmagnesium bromide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Harmful if swallowed.
Reacts violently with water.<b
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-844-1
- Methylmagnesium bromide, 3.2M solution in 2-methyltetrahydrofuran
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P223-Do not allow contact with water.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.