Learn More
Thermo Scientific Chemicals Methylene Blue trihydrate, 95%, pure
121.61 € - 657.27 €
Chemical Identifiers
CAS | 7220-79-3 |
---|---|
Molecular Formula | C16H24ClN3O3S |
Molecular Weight (g/mol) | 373.90 |
MDL Number | MFCD00150008 |
InChI Key | XQAXGZLFSSPBMK-UHFFFAOYSA-M |
Synonym | Basic Blue 9 trihydrate, C.I. 52015 trihydrate |
PubChem CID | 104827 |
SMILES | O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10173030
|
Thermo Scientific Chemicals
126751000 |
100 g | Glass bottle |
121.61 €
100g |
In Stock
Log in to see stock. |
||||
10695172
|
Thermo Scientific Chemicals
126752500 |
250 g | Glass bottle |
203.01 €
250g |
In Stock
Log in to see stock. |
||||
10175253
|
Thermo Scientific Chemicals
126750010 |
1 kg | Glass bottle |
657.27 €
1kg |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7220-79-3 | |
373.90 | |
XQAXGZLFSSPBMK-UHFFFAOYSA-M | |
104827 |
C16H24ClN3O3S | |
MFCD00150008 | |
Basic Blue 9 trihydrate, C.I. 52015 trihydrate | |
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C |
Specifications
100.0°C to 110.0°C | |
95% | |
MFCD00150008 | |
15, 6132 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, insoluble in ether | |
Authentic | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate | |
104827 | |
15% max. (105°C) | |
Pure | |
Crystalline Powder With Metal Luster |
7220-79-3 | |
C16H24ClN3O3S | |
27, 393 | |
Basic Blue 9 trihydrate, C.I. 52015 trihydrate | |
XQAXGZLFSSPBMK-UHFFFAOYSA-M | |
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C | |
373.90 | |
373.89 | |
95% | |
Green | |
Methylene Blue trihydrate, 95% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'rtecsNumber' : SP5740000
- Methylene Blue trihydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Methylene blue trihydrate, Methylene blue
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.