missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue, Redox Indicator, Solid, C.I.52015, Fisher Chemical™
25.60 € - 98.75 €
Chemical Identifiers
CAS | 7220-79-3 |
---|---|
Molecular Formula | C16H24ClN3O3S |
Molecular Weight (g/mol) | 373.90 |
MDL Number | MFCD00150008 |
InChI Key | XQAXGZLFSSPBMK-UHFFFAOYSA-M |
PubChem CID | 104827 |
IUPAC Name | [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate |
SMILES | O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C |
Description
Chemical Identifiers
7220-79-3 | |
373.90 | |
XQAXGZLFSSPBMK-UHFFFAOYSA-M | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate |
C16H24ClN3O3S | |
MFCD00150008 | |
104827 | |
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C |
Specifications
100°C | |
C16H24ClN3O3S | |
XQAXGZLFSSPBMK-UHFFFAOYSA-M | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate | |
104827 | |
Glass Jar | |
3 to 4.5 |
7220-79-3 | |
MFCD00150008 | |
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C | |
373.90 | |
373.89g/mol | |
Green | |
Solid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Methylene Blue trihydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Methylene blue trihydrate, Methylene blue