missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylcyclohexane, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | -126°C |
Color | Colorless |
Density | 0.77g/mL at 25°C |
Boiling Point | 100°C to 102°C (1.013 hPa) |
Flash Point | −4°C (24.8°F) |
Odor | Characteristic |
Packaging | Stainless steel can |
Linear Formula | C6H11CH3 |
Quantity | 20 L |
UN Number | UN2296 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction