missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylamine, Technical, 25/30% (w/v) Solution In Water, Fisher Chemical
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Specifications
Specifications
CAS | 74-89-5 |
Melting Point | -38°C |
Color | Colorless |
pH | 12 |
Boiling Point | 48°C |
Packaging | Amber glass bottle |
Molecular Formula | CH5N |
MDL Number | 8104 |
Quantity | 500 mL |
Synonym | methylamine, aminomethane, monomethylamine, carbinamine, mercurialin, n-methylamine, methylaminen, metilamine, metyloamina, anhydrous methylamine |
Show More |
Safety and Handling
Product Identifier
- Methylamine, solution 25/30% w/v
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction