missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl sulfoxide-d6, for NMR, contains 0.03% TMS, 99.8 atom% D, Thermo Scientific Chemicals
62.91 € - 221.10 €
Chemical Identifiers
CAS | 2206-27-1 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 84.17 |
MDL Number | MFCD00002090 |
InChI Key | IAZDPXIOMUYVGZ-WFGJKAKNSA-N |
Synonym | dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide |
PubChem CID | 75151 |
SMILES | [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10190512
|
Thermo Scientific Acros
360000100 |
10 mL | Glass Bottle |
62.91 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16338196
|
Thermo Scientific Acros
360000250 |
25 mL | Glass Bottle |
132.66 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10059041
|
Thermo Scientific Acros
360000500 |
50 mL | Glass Bottle |
221.10 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
2206-27-1 | |
84.17 | |
IAZDPXIOMUYVGZ-WFGJKAKNSA-N | |
75151 |
C2H6OS | |
MFCD00002090 | |
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide | |
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H] |
Specifications
Liquid | |
1.1900g/mL | |
88°C | |
2.4 mPa.s (20°C) | |
2206-27-1 | |
190.0°C | |
MFCD00002090 | |
13,124; 16,177 | |
IAZDPXIOMUYVGZ-WFGJKAKNSA-N | |
trideuterio(trideuteriomethylsulfinyl)methane | |
75151 | |
NMR |
Colorless | |
99.75% min. | |
1.19 | |
0.03% max. (Coulometric) | |
20.2°C | |
C2H6OS | |
01, IV, 1279 | |
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide | |
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H] | |
84.17 | |
84.18 | |
Methyl sulfoxide-d6, 99.8 atom%D |
Safety and Handling
Product Identifier
- Methyl sulfoxide-d6
RUO – Research Use Only