Learn More
Methyl propiolate, 97%, Thermo Scientific™
74.10 € - 111.00 €
Chemical Identifiers
CAS | 922-67-8 |
---|---|
Molecular Formula | C4H4O2 |
Molecular Weight (g/mol) | 84.074 |
MDL Number | MFCD00008572 |
InChI Key | IMAKHNTVDGLIRY-UHFFFAOYSA-N |
Synonym | methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester |
PubChem CID | 13536 |
IUPAC Name | methyl prop-2-ynoate |
SMILES | COC(=O)C#C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10621765
|
Thermo Scientific Maybridge
SB01817EA |
10 g | Amber Glass Bottle |
74.10 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10527664
|
Thermo Scientific Maybridge
SB01817EB |
25 g | Amber Glass Bottle |
111.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
922-67-8 | |
84.074 | |
IMAKHNTVDGLIRY-UHFFFAOYSA-N | |
13536 | |
COC(=O)C#C |
C4H4O2 | |
MFCD00008572 | |
methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester | |
methyl prop-2-ynoate |
Specifications
922-67-8 | |
0.9400g/mL | |
C4H4O2 | |
MFCD00008572 | |
IMAKHNTVDGLIRY-UHFFFAOYSA-N | |
methyl prop-2-ynoate | |
13536 | |
97% |
97.0 | |
97% | |
HCidentical toCCO2CH3 | |
methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester | |
COC(=O)C#C | |
84.074 | |
84.07 | |
Methyl propiolate |
Safety and Handling
GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.